Key Documents
Z688339
LightSafe centrifuge tubes
capacity 50 mL, conical polypropylene, sterile
About This Item
UNSPSC Code:
41121703
NACRES:
NB.22
material
black tube
cap (attached)
conical polypropylene
sterility
sterile
feature
RCF 10,000 × g
packaging
pack of 50
manufacturer/tradename
Cole Parmer 634435
capacity
50 mL
General description
Disposable polypropylene tubes and caps with recessed lid to prevent leaking. Frosted writing surface allows for permanent identification and labeling. Autoclavable to 122°C and can be stored in freezers to -100°C. RCF values: 15 mL, 8,400 g/50 mL, 10,000 g
Choose from one of the most recent versions:
Certificates of Analysis (COA)