Skip to Content
Merck
All Photos(1)

Key Documents

Z4902

Sigma-Aldrich

ZD 7114 hydrochloride

≥98% (HPLC)

Synonym(s):

Acetamide, 2-[4-[2-[[(2S)-2-hydroxy-3-phenoxypropyl]amino]ethoxy]phenoxy]-N-(2-methoxyethyl)-, ICI D7114

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N2O6 · HCl
CAS Number:
Molecular Weight:
454.94
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -10.7°, c = 1.0 in methanol

color

white to off-white

solubility

H2O: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.COCCNC(=O)COc1ccc(OCCNC[C@H](O)COc2ccccc2)cc1

InChI

1S/C22H30N2O6.ClH/c1-27-13-12-24-22(26)17-30-21-9-7-20(8-10-21)28-14-11-23-15-18(25)16-29-19-5-3-2-4-6-19;/h2-10,18,23,25H,11-17H2,1H3,(H,24,26);1H/t18-;/m0./s1

InChI key

KCAMZVQSGDBGGF-FERBBOLQSA-N

Biochem/physiol Actions

ZD 7114 is a selective β3 adrenergic receptor agonist. β3 adrenoceptors are expressed in adipose tissue and smooth muscle.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

β-Adrenoceptors are activated either by norepinephrine or epinephrine. Learn more about the role of β-adrenoceptors in protein expression, cell signaling, and metabolism research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service