Z329827
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Distilled water, with venting valve (metal ball)
All Photos(1)
About This Item
Recommended Products
material
low-density polyethylene bottle
natural bottle
round bottle
white closure
wide-mouth bottle
feature
label Distilled water
with cap
with venting valve (metal ball)
packaging
pack of 5 ea
manufacturer/tradename
BRAND 144018
capacity
250 mL
General description
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Other Notes
BR144018 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accessory