Skip to Content
Merck
All Photos(1)

Key Documents

Y1521

Sigma-Aldrich

YS-49 monohydrate

≥98% (HPLC)

Synonym(s):

1,2,3,4-Tetrahydro-1-(1-naphthalenylmethyl)-6,7-Isoquinolinediol hydrobromide monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO2 · HBr · H2O
CAS Number:
Molecular Weight:
404.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.Br.Oc1cc2CCNC(Cc3cccc4ccccc34)c2cc1O

InChI

1S/C20H19NO2.BrH.H2O/c22-19-11-15-8-9-21-18(17(15)12-20(19)23)10-14-6-3-5-13-4-1-2-7-16(13)14;;/h1-7,11-12,18,21-23H,8-10H2;1H;1H2

InChI key

QHCUNMLYODPTIU-UHFFFAOYSA-N

Biochem/physiol Actions

YS-49 protects cells from oxidant injury; anti-inflammatory agent; activator of PI3K/Akt signaling. YS-49, an analog of higenamine, induces heme oxygenase (HO-1) in endothelial cells and protects cells from oxidant injury. The compound was reported to inhibit Ang II-stimulated VSMC proliferation through HO-1 production, which inhibits both the JNK pathway and ROS production. A study (JPET) reported that CKD712 (S-YS-49) confer cardiac protection and anti-inflammatory via activation of the PI3K signal pathway. These effects were antagonized by a PI3K inhibitor, wortmannin.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the PKB/Akt page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to PKB/Akt for your research needs.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service