Skip to Content
Merck
All Photos(1)

Documents

T8279

Sigma-Aldrich

Tissue grinder pestle for CD-1

pkg of 2 ea

Synonym(s):

grinding equipment, grinding tools, tissue grinders

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

packaging

pkg of 2 ea

Legal Information

CD-1 is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurine Legroux et al.
Journal of neuroscience methods, 247, 23-31 (2015-03-31)
Flow cytometry is an efficient and powerful technique to characterize and quantify numerous cells. However, the strengths of this technique have not been widely harnessed in neurosciences due to the critical step of CNS tissue preparation into a single cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service