Skip to Content
Merck
All Photos(1)

Key Documents

T5955

Sigma-Aldrich

3,4,5,6-Tetrahydroxyxanthone hydrate

≥98% (HPLC)

Synonym(s):

3,4,5,6-Tetrahydroxy-9H-xanthen-9-one hydrate, MB7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8O6 · xH2O
CAS Number:
Molecular Weight:
260.20 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

color

light brown to brown

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.Oc1ccc2C(=O)c3ccc(O)c(O)c3Oc2c1O

InChI

1S/C13H8O6.H2O/c14-7-3-1-5-9(16)6-2-4-8(15)11(18)13(6)19-12(5)10(7)17;/h1-4,14-15,17-18H;1H2

InChI key

ZFTTXYRPFMYDRT-UHFFFAOYSA-N

Biochem/physiol Actions

3,4,5,6-Tetrahydroxyxanthone is a Na/K-ATPase inihibitor that inhibits pump function without activating the kinase signaling function. It inhibits Na/K ATPase pump action with an affinity comparable to ouabain, but does not alter Na or ATP affinity, is not blocked by potassium, and it does not activate the Src complex or downstream kinases.

Preparation Note

3,4,5,6-Tetrahydroxyxanthone hydrate is soluble in DMSO at a concentration that is greater than 10 mg/ml.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orhan Aktas et al.
The Journal of experimental medicine, 197(6), 725-733 (2003-03-12)
Statins, known as inhibitors of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase, exhibit numerous functions related to inflammation, such as MHC class II down-regulation, interference with T cell adhesion, and induction of apoptosis. Here we demonstrate that both subcutaneous and oral administration
Rodrigo Quezada et al.
Nanomaterials (Basel, Switzerland), 10(6) (2020-06-12)
Type of metal and metal-oxide NPs added to modify Thin-Film Composites Reverse Osmosis Membranes (TFC-RO) can alter their anti-biofouling properties by changing the dissolution process. The development of a mathematical model can facilitate the selection of these NPs. This work
M Armendariz Ontiveros et al.
Materials (Basel, Switzerland), 12(13) (2019-07-03)
The anti-biofouling and desalination properties of thin film composite reverse osmosis membranes (TFC-RO), modified by the incorporation of copper and iron nanoparticles, were compared. Nanoparticles of metallic copper (CuNPs) and an iron crystalline phase mix (Fe and Fe2O3, FeNPs) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service