Skip to Content
Merck
All Photos(1)

Documents

SRP6020

Sigma-Aldrich

VEGF-C from rat

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

VEGFC, Vascular Endothelial Growth Factor-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

18.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

VEGF-C (Vascular Endothelial Growth Factor-C) is a 129 amino acid recombinant protein that lacks the N-terminal and C-terminal extensions and contains only the middle third VEGF homology domain.

Physical form

Sterile filtered and lyophilized with no additives

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 0.1% acetic acid to a concentration of 0.1-1.0 mg/ml

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-Ke Lu et al.
Pharmaceutical biology, 58(1), 1184-1191 (2020-12-01)
Clinically, Pinellia ternata (Thunb.) Breit. (Araceae) (PT) has been widely used in the treatment of atherosclerosis and hyperlipidaemia, but the underlying mechanisms are still not clearly understood. This research was conducted to confirm the mechanism by which PT affects carotid
Takeshi Hisamatsu et al.
Clinical & experimental metastasis, 32(8), 789-798 (2015-10-08)
The aim of this study is to investigate the significance of lymphatic space invasion (LSI) and tumor VEGF-C expression in the lymphatic spread of ovarian cancer. By performing immunostaining using human ovarian cancer specimens, we first investigated the association between
Xue DU et al.
Experimental and therapeutic medicine, 8(5), 1559-1564 (2014-10-08)
The aim of the present study was to investigate the effect of heat shock protein 90 (Hsp90)-specific inhibitor geldanamycin (GA) on the proliferation and apoptosis induced by vascular endothelial growth factor-C (VEGF-C) in cervical cancer cells. HeLa cells (1×10
Joo-Hee Park et al.
PloS one, 9(9), e109055-e109055 (2014-10-01)
MAZ51 is an indolinone-based molecule originally synthesized as a selective inhibitor of vascular endothelial growth factor receptor (VEGFR)-3 tyrosine kinase. This study shows that exposure of two glioma cell lines, rat C6 and human U251MG, to MAZ51 caused dramatic shape
R Sathish Srinivasan et al.
Genes & development, 28(19), 2175-2187 (2014-10-03)
The mammalian lymphatic vasculature is important for returning fluids from the extracellular tissue milieu back to the blood circulation. We showed previously that Prox1 dosage is important for the development of the mammalian lymphatic vasculature. The lack of Prox1 activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service