Skip to Content
Merck
All Photos(2)

Key Documents

SRP5098

Sigma-Aldrich

ULK3, active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZP434C131

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

195-265 nmol/min·mg

mol wt

~51 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ULK3(25989)

General description

The ULK3 (unc-51 like kinase 3) gene is mapped to human chromosome 15q24.1.

Biochem/physiol Actions

ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins. ULK3 participates in cell cycle checkpoint and prevents the progression of cell division prior to complete chromosome separation. Overexpression of ULK3 stimulates autophagy through fibroblast activation.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ULK3 regulates cytokinetic abscission by phosphorylating ESCRT-III proteins
Caballe A, et al.
eLife, 4(10), e06547-e06547 (2015)
Novel genes affecting blood pressure detected via gene-based association analysis
Zhang H,et al.
Genes & Genomics, g3-115 (2015)
The ULK3 Kinase Is Critical for Convergent Control of Cancer-Associated Fibroblast Activation by CSL and GLI
Goruppi , et al.
Testing, 20(10), 2468-2479 (2017)
Alla Maloverjan et al.
Experimental cell research, 316(4), 627-637 (2009-11-03)
The Hedgehog (Hh) signaling pathway plays crucial roles in embryonic development and is implicated in tissue homeostasis maintenance and neurogenesis in adults. Aberrant activation of Hh signaling is associated with various developmental abnormalities and several types of cancer. Genetic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service