Skip to Content
Merck
All Photos(1)

Documents

SRP3060

Sigma-Aldrich

IFN-lambda 2 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

IL-28A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

19.6 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL28A(282616)

General description

IFN lambda-1,-2, and -3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family and Interferons. All three IFN-lambdas uses a distinct receptor system composed of an IFN-lambdaR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defense similar but distinct than the type I Interferons. (Kotenko, SV. et al. Nat Immunology 2003 [1] p. 69-77). Recombinant human IFN-l2 is a 19.6 kDa protein containing 174 amino acid residues.

Biochem/physiol Actions

IFN lambda-1,-2, and -3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family and Interferons. All three IFN-lambdas uses a distinct receptor system composed of an IFN-lambdaR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defense similar but distinct than the type I Interferons. (Kotenko, SV. et al. Nat Immunology 2003 [1] p. 69-77). Recombinant human IFN-l2 is a 19.6 kDa protein containing 174 amino acid residues.

Sequence

PVARLHGALP DARGCHIAQF KSLSPQELQA FKRAKDALEE SLLLKDCRCH SRLFPRTWDL RQLQVRERPM ALEAELALTL KVLEATADTD PALVDVLDQP LHTLHHILSQ FRACIQPQPT AGPRTRGRLH HWLYRLQEAP KKESPGCLEA SVTFNLFRLL TRDLNCVASG DLCV

Physical form

Lyophilized from 10 mM Acetic Acid.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L S Laursen et al.
FEBS letters, 504(1-2), 36-40 (2001-08-28)
Pregnancy-associated plasma protein-A (PAPP-A) has recently been identified as the proteinase responsible for cleavage of insulin-like growth factor binding protein (IGFBP)-4, an inhibitor of IGF action, in several biological fluids. Cleavage of IGFBP-4 by PAPP-A is believed to occur only
Sergei V Kotenko et al.
Nature immunology, 4(1), 69-77 (2002-12-17)
We report here the identification of a ligand-receptor system that, upon engagement, leads to the establishment of an antiviral state. Three closely positioned genes on human chromosome 19 encode distinct but paralogous proteins, which we designate interferon-lambda1 (IFN-lambda1), IFN-lambda2 and
Guowu Hu et al.
The Journal of clinical investigation, 118(3), 1186-1197 (2008-02-09)
Autophagy is a process by which cells recycle cytoplasm and defective organelles during stress situations such as nutrient starvation. It can also be used by host cells as an immune defense mechanism to eliminate infectious pathogens. Here we describe the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service