Skip to Content
Merck
All Photos(1)

Documents

SRP0605

Sigma-Aldrich

Human CD226, FC Fusion, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

DNAX accessory molecule 1 (DNAM-1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

54.7 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD226(10666)

General description

Human CD226 antigen also known as DNAX accessory molecule 1 (DNAM-1), GenBank Accession No. NM_006566, a.a. 19-247 fused to Fc region of human IgG1, this protein is enzymatically biotinylated using AviTagTM technology and expressed in a HEK293 cell expression system. MW = 54.7 kDa (monomer). This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service