Skip to Content
Merck
All Photos(1)

Documents

SML2512

Sigma-Aldrich

Fosfomycin Trometamol

Synonym(s):

Fosfomycin trometamol, Fosfomycin Trometamol Salt, Fosfomycin Tromethamine, Fosfomycin Tromethanol, Tromethamine-Fosfomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H18NO7P
CAS Number:
Molecular Weight:
259.19
UNSPSC Code:
12352200

biological source

Streptomyces sp.

form

powder

storage temp.

2-8°C

InChI

1S/C4H11NO3.C3H7O4P/c5-4(1-6,2-7)3-8;1-2-3(7-2)8(4,5)6/h6-8H,1-3,5H2;2-3H,1H3,(H2,4,5,6)/t;2-,3+/m.0/s1

InChI key

QZJIMDIBFFHQDW-LMLSDSMGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Fosfomycin trometamol (fosfomycin tromethamine is mainly used for the treatment of uncomplicated urinary tract infections (UTIs). Fosfomycin has good in vitro activity against common uropathogens, such as Escherichia coli (including extended-spectrum β-lactamase-producing E. coli), Proteus mirabilis, Klebsiella pneumoniae and Staphylococcus saprophyticus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service