Skip to Content
Merck
All Photos(1)

Key Documents

SCP0163

Sigma-Aldrich

H.I.V.-1 Protease Substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H58N10O12
Molecular Weight:
846.93
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥88%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH2

Application

Acetyl-SQNYPVV-NH2 is used as a substrate of human immunodeficiency virus-1 (HIV-1) protease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service