SAB5700402
Anti-Phospho-Smad2-S465/467 + Smad3-S423/425 antibody produced in rabbit
Synonym(s):
HSPC193, HsT17436, JV15-2, LDS1C, LDS3, MADH3
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
25
35
43
48
52
species reactivity
rat, human, mouse
concentration
2.74 mg/ml
technique(s)
immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
phosphorylation ((pSer465/pSer467),(pSer423/pSer425))
Gene Information
human ... SMAD2(4087)
Immunogen
Physical form
Storage and Stability
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service