Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300039

Sigma-Aldrich

Anti-phospho-STAT6 (pThr645) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-D12S1644 antibody produced in rabbit, Anti-IL-4-STAT antibody produced in rabbit, Anti-STAT6B antibody produced in rabbit, Anti-STAT6C antibody produced in rabbit, Anti-signal transducer and activator of transcription 6, interleukin-4 induced antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~110 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100

isotype

IgG

immunogen sequence

(P-A-TP-I-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr645)

Gene Information

human ... STAT6(6778)

Immunogen

Peptide sequence around phosphorylation site of threonine 645 (P-A-T(p)-I-K), according to the protein STAT6.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Carries out a dual function: signal transduction and activation of transcription. Involved in interleukin-4 signalling.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service