Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900095

Sigma-Aldrich

Anti-GRK5 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

g protein-coupled receptor kinase 5, g protein-coupled receptor kinase grk5, gprk5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRK5(2869)

Immunogen

Synthetic 15 amino acid peptide from N-terminus of human GRK5. Percent identity with other species by BLAST analysis: Human (100%); Gorilla, Orangutan, Gibbon, Marmoset, Bovine, Horse (93%); Dog, Elephant, Panda, Pig (87%); Mouse, Rat, Bat (80%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service