Skip to Content
Merck
All Photos(1)

Documents

SAB2100686

Sigma-Aldrich

Anti-EPAS1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Endothelial PAS domain protein 1, Anti-HIF2A, Anti-HLF, Anti-MOP2, Anti-PASD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

90 kDa

species reactivity

human, rabbit, horse, bovine, rat, dog, guinea pig, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPAS1(2034)

Immunogen

Synthetic peptide directed towards the middle region of human EPAS1

Biochem/physiol Actions

EPAS1 is a transcription factor involved in the induction of oxygen regulated genes. EPAS1 binds to core DNA sequence 5′-[AG]CGTG-3′ within the hypoxia response element (HRE) of target gene promoters. EPAS1 regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of lung. EPAS1 may also play a role in the formation of the endothelium that gives rise to the blood brain barrier. EPAS1 is a potent activator of the Tie-2 tyrosine kinase expression. The activation seems to require recruitment of transcriptional coactivators such as CREBPB and probably EP300. Interaction of EPAS1 with redox regulatory protein APEX seems to activate CTAD.

Sequence

Synthetic peptide located within the following region: ESLFKPHLMAMNSIFDSSGKGAVSEKSNFLFTKLKEEPEELAQLAPTPGD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service