Skip to Content
Merck
All Photos(2)

Key Documents

S2069

Sigma-Aldrich

Anti-SNAP-29 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Synaptosomal-Associated Protein 29

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 29 kDa

species reactivity

mouse, rat

technique(s)

indirect immunofluorescence: 1:100 using mouse fibroblast NIH3T3 cell line.
microarray: suitable
western blot: 1:1,000 using whole cell extract of mouse fibroblasts NIH3T3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Synaptosomal-associated protein 29 kDa (SNAP-29), is a non-neuronal isoform of synaptosomal-associated protein 25 (SNAP-25), originally isolated from a human cDNA library in a yeast two-hybrid screen for proteins interacting with syntaxin 3. It is localized predominantly in intracellular membrane structures specifically with the Golgi apparatus. SNAP-29 is ubiquitously expressed, in neuronal and non-neuronal cells. It codes for a protein which belongs to the N-ethylmale-imide-sensitive factor-attachment protein receptor (SNARE) family of proteins. SNAP-29 gene is mapped to human chromosome 22q11.2.

Specificity

This sequence is identical in rat SNAP-29 and highly conserved (single amino acid substitution) in human SNAP-29.

Immunogen

synthetic peptide corresponding to the C-terminus of mouse SNAP-29 (amino acids 238-257), conjugated to KLH.

Application

Anti-SNAP-29 antibody produced in rabbit may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Synaptosomal-associated protein 29 kDa (SNAP-29) plays an important role in intra-Golgi traffic and regulates a variety of intracellular membrane trafficking pathways. SNAP-29 is capable of participating in various intracellular transport steps, by interacting with different syntaxins and vesicle-associated membrane proteins (VAMPs), specifically localized to different membrane compartments. It is present in synapses and is capable of interacting with syntaxin 1A. For binding to the N-ethylmale-imide-sensitive factor-attachment protein receptor (SNARE) complex, SNAP-29 competes with α-SNAP and subsequently controls synaptic transmission by preventing disassembly of the SNARE complex. It plays a role in the endocytosis of insulin-like growth factor 1 (IGF-1) receptors, by directly interacting with insulin like growth factor1 receptor (IGF1R). Bi-allelic mutations in SNAP-29 is associated with cerebral dysgenesis, neuropathy, ichthyosis, and keratoderma (CEDNIK) syndrome. In infants, deletion of chromosome 22q11.2 results in DiGeorge and Velocardial facial syndrome (22q11.2DS).

Target description

SNAP-29 , a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of SNAP29 impairs endocytic recycling and cell motility
Rapaport D, et al.
PLoS ONE, 5(3), e9759-e9759 (2010)
Irini Topalidou et al.
PLoS genetics, 12(5), e1006074-e1006074 (2016-05-19)
The dense-core vesicle is a secretory organelle that mediates the regulated release of peptide hormones, growth factors, and biogenic amines. Dense-core vesicles originate from the trans-Golgi of neurons and neuroendocrine cells, but it is unclear how this specialized organelle is
The last exon of SNAP-23 regulates granule exocytosis from mast cells
Vaidyanathan VV, et al.
The Journal of Biological Chemistry, 276(27), 25101-25106 (2001)
Evidence-based guidelines for controlling pH in mammalian live-cell culture systems
Michl J, et al.
Communications biology, 2(1), 1-12 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service