description
non-ionic
SMILES string
CCCCCCCCCCCCCCCCCC(=O)OC[C@@H](O)[C@H]1OC[C@H](OC(=O)CCCCCCCCCCCCCCCCC)[C@H]1OC(=O)CCCCCCCCCCCCCCCCC
Other Notes
Fatty acid composition:
stearic acid (C18:0) approx. 50%; balance primarily palmitic acid (C16:0).
stearic acid (C18:0) approx. 50%; balance primarily palmitic acid (C16:0).
Legal Information
Span is a registered trademark of Croda International PLC
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
302.0 °F - closed cup
Flash Point(C)
150.00 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service