Skip to Content
Merck
All Photos(1)

Documents

P4896

Sigma-Aldrich

Pararosaniline acetate

Synonym(s):

Basic Parafuchsin, Basic Red 9, Magenta O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3 · C2H4O2
CAS Number:
Molecular Weight:
347.41
Colour Index Number:
42500
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

203 °C (dec.) (lit.)

SMILES string

CC(O)=O.Nc1ccc(cc1)C(\c2ccc(N)cc2)=C3\C=CC(=N)C=C3

InChI

1S/C19H17N3.C2H4O2/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;1-2(3)4/h1-12,20H,21-22H2;1H3,(H,3,4)

InChI key

YIXIVOYGLPFDCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This is the pure, principal component of mixtures commonly referred to as Basic Fuchsin.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service