MTCR8014
MTC™ Bio IceRack™ Tube Platform with Detachable Lid
to hold, 100 x 1.5 mL tubes
Synonym(s):
cooled microtube rack, cooled tube rack
All Photos(1)
About This Item
Recommended Products
material
polypropylene
feature
to hold 100 x 1.5 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
MTC™ Bio R8014
parameter
-80-135 °C temp. range
Related Categories
General description
IceRack™ boxes have a removable white tube tray that allows the blue base to be filled with crushed ice or other temperature-controlling materials. They also feature a translucent cover. Temperature-sensitive samples and assays in microtubes can be processed with an extra degree of temperature stability. These boxes are suitable for use with screw-cap or snap-cap microtubes from 0.5mL to 1.5/2.0mL.
IceRack™ for 100 × 1.5mL tubes, detachable lid, tube platform, and base, numbered cavities, accepts snap cap and screw cap 1.5mL to 2.0mL tubes, 6/cs
Features and Benefits
- Designed for crushed Ice
- Polypropylene, -80°C to 135°C
- Detachable “lift-off” lids
- Economical
- For screw cap and snap cap tubes
Legal Information
IceRack is a trademark of MTC Bio
MTC is a trademark of MTC Bio