Skip to Content
Merck
All Photos(2)

Key Documents

M4295

Sigma-Aldrich

Maltotriitol

≥95%

Synonym(s):

α-D-Glc-(1→4)-α-D-Glc-(1→4)-D-glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O16
CAS Number:
Molecular Weight:
506.45
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

OCC(O)C(O)C(OC1OC(CO)C(OC2OC(CO)C(O)C(O)C2O)C(O)C1O)C(O)CO

InChI

1S/C18H34O16/c19-1-5(23)9(25)15(6(24)2-20)33-18-14(30)12(28)16(8(4-22)32-18)34-17-13(29)11(27)10(26)7(3-21)31-17/h5-30H,1-4H2

InChI key

XJCCHWKNFMUJFE-UHFFFAOYSA-N

General description

Maltotriitol is a sugar.

Application

The presence of maltotriitol (C(18)H(34)O(16)) in maltose syrup is responsible for a change of the crystal morphology in the industrial crystallization process of maltitol (C(12)H(24)O(11)). Maltotriitol can be used as an inhibitor of acid production in human dental plaque.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Aizawa et al.
Caries research, 43(1), 17-24 (2009-01-13)
This study evaluated acid production from cooked starch by Streptococcus mutans, Streptococcus sobrinus, Streptococcus sanguinis and Streptococcus mitis, and the effects of alpha-amylase inhibitors (maltotriitol and acarbose) and xylitol on acid production. Streptococcal cell suspensions were anaerobically incubated with various
Maltotriitol inhibition of maltose metabolism in Streptococcus mutans via maltose transport, amylomaltase and phospho-alpha-glucosidase activities.
P Würsch et al.
Caries research, 19(5), 439-449 (1985-01-01)
H Kondo et al.
Carbohydrate research, 206(1), 161-166 (1990-09-30)
The effect of the oligosaccharide analog maltotriitol (G3OH) on the action pattern of porcine pancreatic alpha-amylase (PPA) was examined using amylose as a substrate. Fluorescence titration indicated that two molecules of G3OH can bind to one molecule of PPA. The
Inhibition of human digestive enzymes by hydrogenated malto-oligosaccharides
Wursch, P. and S. Vedovo
Internat. J. Vit. Nutr. Res., 51, 161-165 (1981)
Maltitol and maltotriitol as inhibitors of acid production in human dental plaque.
P Würsch et al.
Caries research, 16(2), 90-95 (1982-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service