Skip to Content
Merck
All Photos(1)

Key Documents

K3894

Sigma-Aldrich

KMUP-1

≥98% (HPLC)

Synonym(s):

1H-Purine-2,6-dione, 7-[2-[4-(2-chlorophenyl)-1-piperazinyl]ethyl]-3,7-dihydro-1,3-dimethyl-, 7-[2-[4-(2-Chlorophenyl)piperazinyl]ethyl]-1,3-dimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23ClN6O2
CAS Number:
Molecular Weight:
402.88
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

CN1C(=O)N(C)c2ncn(CCN3CCN(CC3)c4ccccc4Cl)c2C1=O

Biochem/physiol Actions

KMUP-1 is a soluble guanylyl cyclase activator and PDE inhibitor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service