Skip to Content
Merck
All Photos(1)

Key Documents

K1013

Sigma-Aldrich

Kuanoniamine C from Oceanappia sp.

97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18N4OS
CAS Number:
Molecular Weight:
374.46
MDL number:
UNSPSC Code:
12352202

Assay

97% (HPLC)

storage temp.

−20°C

SMILES string

CCCNCCc1cccc2c3ccnc4c5ncsc5cc([nH]c12)c34

Biochem/physiol Actions

Inhibits proliferation of select cell types; binds to adenosine receptor subtypes A1 and A2A.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q Ding et al.
Current medicinal chemistry, 6(1), 1-27 (1999-01-05)
Marine organisms are a rich source for natural products. Pyrrolo[4,3, 2-de]quinolines and pyrido[4,3,2-mn]acridines are of major interest as metabolites in sponges and ascidians. Many of these compounds have generated interest both as challenging problems for structure elucidation and synthesis as
C Eder et al.
Journal of natural products, 61(2), 301-305 (1998-03-26)
The Micronesian sponge Oceanapia sp. afforded three pyridoacridine alkaloids: the known compounds kuanoniamine C (1) and kuanoniamine D (2), as well as the new N-deacyl derivative (3) of the kuanoniamines. Compounds 1 and 2 exhibited insecticidal activity toward neonate larvae

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service