Skip to Content
Merck
All Photos(1)

Documents

H7786

Sigma-Aldrich

Hck human

recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in baculovirus infected insect cells

Quality Level

form

buffered aqueous glycerol solution

specific activity

≥350 units/mg protein

mol wt

58 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HCK(3055)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hck is a member of the Src family of non-receptor associated protein tyrosine kinases (PTK).

Unit Definition

One unit will transfer 1 nanomole of phosphate per minute using polyE4Y as the substrate at pH 7.5 at 30 °C

Physical form

Solution in 50 mM Tris pH 7.5, containing 0.05 mM EDTA, 1mM DTT, 100 mM NaCl, 0.05% NP-40, and 50% glycerol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine Astarie-Dequeker et al.
Journal of cell science, 115(Pt 1), 81-89 (2002-01-22)
In macrophages, lysosomes are suspected to have a heterogenous population of vesicles. This study was thus undertaken to identify and to characterize lysosomal compartments in human macrophages. Hck is a Src-family tyrosine kinase associated with secretory lysosomes in neutrophils and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service