Skip to Content
Merck
All Photos(1)

Documents

G211040

Sigma-Aldrich

DMT-2′O-TBDMS-rG(ib) Phosphoramidite

configured for MerMade

Synonym(s):

N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H68N7O9PSi
CAS Number:
Molecular Weight:
970.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for MerMade

nucleoside profile

base: guanosine
base protecting group: isobutyryl
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: standard

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C50H68N7O9PSi/c1-32(2)45(58)54-48-53-44-41(46(59)55-48)52-31-56(44)47-43(66-68(12,13)49(7,8)9)42(65-67(63-29-17-28-51)57(33(3)4)34(5)6)40(64-47)30-62-50(35-18-15-14-16-19-35,36-20-24-38(60-10)25-21-36)37-22-26-39(61-11)27-23-37/h14-16,18-27,31-34,40,42-43,47H,17,29-30H2,1-13H3,(H2,53,54,55,58,59)/t40-,42-,43-,47-,67?/m1/s1

InChI key

PGJKESPHANLWME-FTZVBZPOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

RNA phosphoramidites, along with RNA CPG, and analogous to DNA synthesis support the synthesis of sequence-specific RNA oligonucleotides for RNA interference (RNAi) and can be used in target validation and other drug development techniques.Modern phosphoramidite-mediated synthesis has enabled routine high-yielding preparations of RNA oligonucleotides. High-quality RNA phosphoramidites are key to low failure rates, the high biological activity of synthesis products, and cost-effectiveness.Key Features
  • Industry-standard 2′O-TBDMS protective group
  • Consistent lot-to-lot purity and performance
  • Compatible with deprotection methods based on methylamine or AMA
  • Standard RNA phosphoramidites provide excellent coupling results whenused with ETT or BTT as an activator; best results are obtained withActivator 42®
  • Capping with standard acetic anhydride capping reagent rather than withFast Deprotection Cap A
  • Manufactured under a certified ISO 9001 quality systemDMT-2′O-TBDMS-rG(ib)
Phosphoramidite is configured for MerMade Synthesizers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service