Skip to Content
Merck
All Photos(1)

Key Documents

F2144

Sigma-Aldrich

1,1′-Ferrocenedicarboxylic acid

~96%

Synonym(s):

1,1′-Biscarboxylferrocene, 1,1′-Dicarboxyferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10FeO4
CAS Number:
Molecular Weight:
274.05
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

~96%

mp

≥300 °C (lit.)

SMILES string

[Fe].OC(=O)[C]1[CH][CH][CH][CH]1.OC(=O)[C]2[CH][CH][CH][CH]2

InChI

1S/2C6H5O2.Fe/c2*7-6(8)5-3-1-2-4-5;/h2*1-4H,(H,7,8);

InChI key

ISVVAJYTLASNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Planar chiral ferrocene-based amido-phosphine ligands as catalysts for asymmetric allylic alkylation reactions
  • Potential antitumor agents
  • Isostructural ferrocenyl coordination polymer hollow microspheres with H2 storage properties
  • Prganometallic aromatic polyesters
  • Manganese-lanthanide-ferrocene clusters

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service