F1274
Carestream® X-Omat LS film
Synonym(s):
autoradiography film, xray film
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105341
NACRES:
NB.22
Recommended Products
size
5 in. (13 cm) × 7 in. (18 cm)
Looking for similar products? Visit Product Comparison Guide
General description
Kodak Life Science Imaging film is equivalent to XRP. Coated with emulsion on both sides of a blue-tinted base, produces higher quality images than XAR film (improved definition with less background fog), however, the exposure time is approximately doubled. Sheets of film are interleaved with sheets of paper.
Legal Information
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Soo Jin Jeon et al.
PloS one, 9(3), e92723-e92723 (2014-03-25)
The emergence of antibiotic resistant microorganisms is a great public health concern and has triggered an urgent need to develop alternative antibiotics. Chitosan microparticles (CM), derived from chitosan, have been shown to reduce E. coli O157:H7 shedding in a cattle
Fernando Moreira Simabuco et al.
Journal of cellular biochemistry, 120(4), 5597-5611 (2018-10-16)
In a previous study, we have shown that the gene promoter of a protein termed KIAA0082 is regulated by interferon and that this protein interacts with the RNA polymerase II. It has been subsequently shown that KIAA0082 is the human
Minzi Deng et al.
Experimental cell research, 395(1), 112175-112175 (2020-07-18)
Autophagy is a basic catabolic response that eukaryotic cells use to degrade unnecessary or dysfunctional cellular components in an orderly and regulated manner. It plays important roles in maintaining cellular homeostasis, energy homeostasis, response to environmental stimuli, and the development
Long-term, hormone-responsive organoid cultures of human endometrium in a chemically defined medium.
Margherita Y Turco et al.
Nature cell biology, 19(5), 568-577 (2017-04-11)
In humans, the endometrium, the uterine mucosal lining, undergoes dynamic changes throughout the menstrual cycle and pregnancy. Despite the importance of the endometrium as the site of implantation and nutritional support for the conceptus, there are no long-term culture systems
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service