EP6001071910
Eppendorf® Side Access Stainless Steel Rack for CryoCube(R) F740 Upright Freezers
including front and back handle for storage of 24 boxes up to 136x136x63 mm (2.5 inch boxes), 3-Compartment, external W × D × H (139 mm) (569 mm) (406 mm)
About This Item
Recommended Products
material
stainless steel
feature
3-Compartment
Holds 24 x boxes (2.5 in.)
including front and back handle for storage of 24 boxes up to 136x136x63 mm (2.5 inch boxes)
manufacturer/tradename
Eppendorf® 6001071910
technique(s)
cryopreservation: suitable
external W × D × H
(139 mm) (569 mm) (406 mm)
size
549x139x444 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service