Documents
EP5895130003
Rotor for Eppendorf® 5920R Centrifuges
FA-20x5, aerosol tight, to hold, 20 x 5.0 mL Eppendorf tubes, with Eppendorf QuickLock® rotor lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 20 x 5.0 mL Eppendorf tubes
aerosol tight
fixed angle rotor
with Eppendorf QuickLock® rotor lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895130003 (Global)
parameter
25,062 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5920R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG