Skip to Content
Merck
All Photos(1)

Key Documents

EHU004071

Sigma-Aldrich

MISSION® esiRNA

targeting human CYP2E1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACGTCATAGCCGACATCCTCTTCCGCAAGCATTTTGACTACAATGATGAGAAGTTTCTAAGGCTGATGTATTTGTTTAATGAGAACTTCCACCTACTCAGCACTCCCTGGCTCCAGCTTTACAATAATTTTCCCAGCTTTCTACACTACTTGCCTGGAAGCCACAGAAAAGTCATAAAAAATGTGGCTGAAGTAAAAGAGTATGTGTCTGAAAGGGTGAAGGAGCACCATCAATCTCTGGACCCCAACTGTCCCCGGGACCTCACCGACTGCCTGCTCGTGGAAATGGAGAAGGAAAAGCACAGTGCAGAGCGCTTGTACACAATGGACGGTATCACCGTGACTGTGGCCGACCTGTTCTTTGCGGGGACAGAGACCACCAGCACAACTCTGAGATATGGGCTCCTGATTCTCATGAAATACCCTGAGATTGAAGAGAAGCTCCATGAAGAAATTGACAGGGTGATTGGGCCAAGCCGAATCCCTGCCATCAAGGATAGGCAAGAGATGCCCTACATGGATGCTGTGGTGCATGAGATTCAGCGGTTCATCACC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshio Ijiri et al.
Xenobiotica; the fate of foreign compounds in biological systems, 48(1), 60-72 (2017-01-14)
1. Drug-induced liver injury is difficult to predict at the pre-clinical stage. This study aimed to clarify the roles of caspase-8 and -9 in CYP2E1 metabolite-induced liver injury in both rats and cell cultures in vitro treated with carbon tetrachloride (CCl
Beomseok Son et al.
American journal of physiology. Lung cellular and molecular physiology, 313(5), L916-L929 (2017-08-12)
Radiation-induced pulmonary fibrosis (RIPF) is one of the most common side effects of lung cancer radiotherapy. This study was conducted to identify the molecular mechanism responsible for RIPF. We revealed that the transcriptional level of cytochrome P450 2E1 (CYP2E1) was
Lu Li et al.
International journal of oncology, 50(5), 1832-1838 (2017-03-25)
Although vitamin K2 (VK2) exhibits inhibitory effects on the viability of hepatoma cells, hepatoma cells are insensitive to VK2. Therefore, this investigation is an attempt to enhance the sensitivity of hepatoma cells to VK2. Our results showed that VK2 acted
Liliana Mancio-Silva et al.
Cell metabolism, 29(3), 727-735 (2019-03-07)
The liver plays a central role in metabolism; however, xenobiotic metabolism variations between human hepatocytes and those in model organisms create challenges in establishing functional test beds to detect the potential drug toxicity and efficacy of candidate small molecules. In
Dahlia Triningsih et al.
Toxicology in vitro : an international journal published in association with BIBRA, 72, 105105-105105 (2021-02-06)
Acrylamide is known as a neurotoxicant found in commonly consumed food as well as in human body. However, the underlying mechanisms involved in neurotoxicity by acrylamide and its metabolite, glycidamide remain largely unknown. In this study, we have examined the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service