E2275
pFLAG-CMV™−6a,b,c Expression Vectors
set of pFLAG-CMV-6a, -6b, and -6c vectors for cytoplasmic expression of N-terminal FLAG
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
MDL number:
UNSPSC Code:
12352200
Recommended Products
tag
FLAG® tagged
grade
for molecular biology
form
buffered aqueous solution
shipped in
dry ice
storage temp.
−20°C
General description
p-FLAG-CMV-6a, p-FLAG-CMV-6b, and p-FLAG-CMV-6c Expression Vectors used for the expression of N-terminal FLAG® tagged proteins in mammalian cells. pFLAG-CMV-6 a,b,c Expression Vectors are used as shuttle vectors for E. coli and mammalian cells. These vectors facilitate the intracellular expression of N-terminal FLAG-tagged fusion proteins. Transient expression in mammalian cells is driven by the CMV promoter-regulatory region. The SV40 origin of replication results in most efficient replication in mammalian cells expressing the SV40 large T antigen. The a, b, and c vector series makes all three reading frames available for every restriction site in the MCS.
Vector Maps and Sequences
Vector Maps and Sequences
Application
p-FLAG-CMV-6a, p-FLAG-CMV-6b, and p-FLAG-CMV-6c Expression Vectors are suitable for cytoplasmic expression of N-terminal FLAG® tagged proteins.
Components
- p-FLAG-CMV-6a Expression Vector (20μg)
- p-FLAG-CMV-6b Expression Vector (20 μg)
- p-FLAG-CMV-6c Expression Vector (20 μg)
- pFLAG-CMV-6a-BAP control plasmid (20 μg)
Legal Information
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC
related product
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Virginie W Gautier et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(45), 16362-16367 (2005-11-02)
The primary function of the HIV-1 regulatory protein Tat, activation of transcription from the viral LTR, is highly regulated by complex interactions between Tat and a number of host cell proteins. Tat nuclear import, a process mediated by importin beta
Rick F Nelson et al.
The Journal of biological chemistry, 281(29), 20242-20251 (2006-05-10)
In SCF (Skp1/Cullin/F-box protein) ubiquitin ligases, substrate specificity is conferred by a diverse array of F-box proteins. Only in fully assembled SCF complexes, it is believed, can substrates bound to F-box proteins become ubiquitinated. Here we show that Fbx2, a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service