Skip to Content
Merck
All Photos(1)

Documents

D9439

Sigma-Aldrich

Desmosine chloride

from bovine, 99.5%±0.2%

Synonym(s):

DES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40ClN5O8
CAS Number:
Molecular Weight:
562.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.56

biological source

bovine

Quality Level

Assay

99.5%±0.2%

form

lyophilized powder

purified by

chromatography (and purified by solvent extraction)

storage temp.

2-8°C

SMILES string

[Cl-].NC(CCCC[n+]1cc(CCC(N)C(O)=O)c(CCCC(N)C(O)=O)c(CCC(N)C(O)=O)c1)C(O)=O

InChI

1S/C24H39N5O8.ClH/c25-17(21(30)31)5-1-2-11-29-12-14(7-9-19(27)23(34)35)16(4-3-6-18(26)22(32)33)15(13-29)8-10-20(28)24(36)37;/h12-13,17-20H,1-11,25-28H2,(H3-,30,31,32,33,34,35,36,37);1H

InChI key

FUPMLZCXPJSGPV-UHFFFAOYSA-N

General description

Each vial contains 1 μmole of purified desmosine.

Application

Desmosine chloride is often used in assays for free or urinary desmosine as markers for disease states characterized by elastin degradation (e.g., emphysema). Elastin degradation has shown strong correlation with vascular calcification, and pretreatment with aluminum ions can inhibit the binding of matrix metalloproteinase (MMP) to elastin.
Assays for free or urinary desmosine are often used as markers for disease states characterized by elastin degradation (e.g., emphysema). Conversely, excessive levels of incorporated desmosine are indicators of fibrotic conditions (e.g., cirrhosis).

Biochem/physiol Actions

Desmosine is a polyfunctional amino acid that forms cross-links in mature elastin.

Caution

Extremely hygroscopic

Analysis Note

Calibrated by UV and amino acid analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franca Cocci et al.
The international journal of biochemistry & cell biology, 34(6), 594-604 (2002-04-12)
An enhanced proteolysis of lung interstitium is key event in the pathogenesis of emphysema, a major constituent of chronic obstructive pulmonary disease. To assess whether urinary desmosine and/or hydroxyproline may be used as a marker of lung destruction we studied
M Bailey et al.
The American journal of pathology, 159(6), 1981-1986 (2001-12-26)
Calcification of elastin occurs in many pathological cardiovascular diseases including atherosclerosis. We have previously shown that purified elastin when subdermally implanted in rats undergoes severe calcification and aluminum chloride (AlCl(3)) pretreatment of elastin inhibits calcification. In the present study we
Dina M Basalyga et al.
Circulation, 110(22), 3480-3487 (2004-11-17)
Elastin calcification is a widespread feature of vascular pathology, and circumstantial evidence exists for a correlation between elastin degradation and calcification. We hypothesized that matrix metalloproteinase (MMP)-mediated vascular remodeling plays a significant role in elastin calcification. In the present studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service