Skip to Content
Merck
All Photos(1)

Documents

MAT-Lu

94102735, rat prostate, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MAT-Lu, 94102735

biological source

rat prostate

growth mode

Adherent

karyotype

Modal no. 85

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat dorsal prostate adenocarcinoma

Cell Line Description

MAT-Lu is one of a large series of cell lines of transplantable prostatic tumours originating from the spontaneous Dunning R-3327 rat prostatic cancer. The MAT-Lu cell line has been derived from a Metastatic-Anaplastic Tumour metastasising to the lungs (=MAT-Lu) and is androgen-independent.

Application

Carcinogenesis studies, cytogenetic analysis

Culture Medium

RPMI 1640 + 2mM Glutamine + 250nM Dexamethasone (DXMT) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 2-4x1000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service