Skip to Content
Merck
All Photos(1)

Documents

HAM 006

from human blood, 94082275

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

human blood

description

Human HLA-Typed Lymphoblastoid Cell Line

growth mode

Suspension

morphology

Spherical

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

An Epstein-Barr virus (EBV) transformed lymphoblastoid cell line with Human Leukocyte Antigen (HLA)-typing data available.

Subculture Routine

RPMI 1640 (R0883) + 2mM L-Glutamine (G7513) + 10% FBS (F2442). To help establish a culture the concentration of FBS can be increased to 20%.
When resuscitating from frozen, cells should be seeded at 0.5 x 106 cells/ml in the culture medium. The culture should establish log phase growth after 3-5 days. When growing well dilute every 1-3 days to 3 x 105 cells/ml. Maintain cultures between 3-9 x 105 cells/ml for maximum growth; 5% CO2; 37°C. Cells form floating aggregates of varying size.

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Citation

If use of this culture results in a scientific publication, it should be cited in the publication as: HAM 006 (ECACC 94082275)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service