biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
Sp2/0
Reactivity
Anti-IgG2a, mouse
technique(s)
cell culture | mammalian: suitable
isotype
IgM
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse Sp2/0 x Mouse BALB/c spleen
Cell Line Description
The immunisation protocol involved repeated intranasal infection with a single i.v. injection before fusion. The IgM antibodies bind murine IgG2a of "a" allotype and Fc region. There is no cross-reactivity with A/PR/8/34 influenza virus, DNP, NP, DNA or cytoskeletal proteins. Contains a CH2 domain.
Immunogen
A/PR/8/34 mouse pathogenic human influenza virus
Application
Differential detection of complexed IgG2a in the presence of native IgG2a, allotype detection or quantitation.
Culture Medium
RPMI 1640 or DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS) (can be adapted to 1% Foetal Bovine Serum (FBS))
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service