Skip to Content
Merck
All Photos(1)

Key Documents

7D4

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity: Anti-Interleukin-2 receptors, murine, 88111402

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Rat Heterohybridoma

packaging

tube of 5 μg 88111402-DNA-5UG
pkg of vial of cells 88111402-1VL

ImmunoDon

Lewis Rat spleen

Myeloma

Sp2/0

Reactivity

Anti-Interleukin-2 receptors, murine

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Rat Lewis spleen

Cell Line Description

The antibody is directed against the murine IL-2 receptor ligand complex present on activated murine T and B cells. It inhibits proliferation of activated cells in the presence of IL-2, but does not block the binding of IL-2.

Immunogen

Murine IL-2 dependent HT-2 cells

Application

The isotype of the monoclonal antibody produced by this cell line is rat IgM, kappa.

Culture Medium

DMEM + 2mM glutamine + 1% Non Essential Amino Acids (NEAA) + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service