Skip to Content
Merck
All Photos(1)

Documents

H9C2 (2-1) Cell Line from rat

88092904, rat cardiovascular , Myoblast

Synonym(s):

H9c2 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

H9C2 (2-1) Cell Line from rat, 88092904, BDIX heart myoblast

biological source

rat cardiovascular

growth mode

Adherent

karyotype

Not specified

morphology

Myoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat BDIX heart myoblast

Cell Line Description

Skeletal muscle properties. Myotubes formed at confluency respond to acetylcholine. Differentiation improved by reducing serum concentration to 1%.

Application

Differentiation studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Important to passage at sub-confluence, reclone periodically to keep differentiated characteristics. Cells will fuse to form myotubes if allowed to reach confluency.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service