Skip to Content
Merck
All Photos(1)

Key Documents

C9477

Sigma-Aldrich

Anti-Chloride Channel-2 (CLC-2, Clcn2) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot (chemiluminescent): 1:200-1:400 using rat membranes and ECL

UniProt accession no.

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide corresponding to amino acids 888-906 of the rat chloride channel-2 (CLC-2).

Application

Anti-Chloride Channel-2 antibody produced in rabbit is suitable for western blot (chemiluminescent) at a working dilution of 1:200-1:400 using rat membranes and ECL.

Biochem/physiol Actions

The encoded protein, voltage-gated chloride channel, is a transmembrane protein that maintains chloride ion homeostasis in various cells. Defects in this gene may be a cause of certain epilepsies. Autosomal-recessive mutations ion this gene cause a leukoencephalopathy that affects brain ion and water homoeostasis. It is characterized by intramyelinic oedema. It is involved in maintaining the volume of trabecular meshwork cells by adjusting the outflow of aqueous solutions and for maintaining the fluid balance. It may be a potential target for the treatment of primary open angle glaucoma.

Physical form

Lyophilized at ~0.6 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Wei Wang et al.
Molecular medicine reports, 8(4), 1099-1105 (2013-08-13)
The chloride channel protein 2 (CLC‑2) is important in maintaining the volume of trabecular meshwork cells by adjusting the outflow of aqueous solutions and maintaining the fluid balance. However, little is known concerning the functions of CLC‑2 in the cytoskeleton
Cécile Saint-Martin et al.
Human mutation, 30(3), 397-405 (2009-02-05)
Heterozygous mutations in the CLCN2 gene encoding the voltage-gated chloride channel CLC2 have been identified in patients with idiopathic generalized epilepsy (IGE). Yet the involvement of CLCN2 in epilepsy remains controversial. To investigate the involvement of CLCN2 in another independent
Christel Depienne et al.
The Lancet. Neurology, 12(7), 659-668 (2013-05-28)
Mutant mouse models suggest that the chloride channel ClC-2 has functions in ion and water homoeostasis, but this has not been confirmed in human beings. We aimed to define novel disorders characterised by distinct patterns of MRI abnormalities in patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service