C213250
DMT-2′Fluoro-dC(ac) Phosphoramidite
configured for (ÄKTA® and OligoPilot®)
Synonym(s):
DMT-2′-F-dC(ac) amidite
About This Item
Recommended Products
biological source
non-animal source (no BSE/TSE risk)
Quality Level
product line
Proligo Reagents
Assay
≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)
form
powder
technique(s)
oligo synthesis: suitable
impurities
≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content
color
white to off-white
λ
conforms (UV/VIS Identity)
suitability
conforms to structure for H-NMR
conforms to structure for LC-MS
compatibility
configured for (ÄKTA® and OligoPilot®)
nucleoside profile
base: deoxycytidine
base protecting group: acetyl
2' protecting group: fluoro
5' protecting group: DMT
deprotection: fast/standard
storage temp.
−20°C
SMILES string
COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](F)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(NC(C)=O)=NC3=O)(c4ccccc4)c5ccc(OC)cc5
InChI
1S/C41H49FN5O8P/c1-27(2)47(28(3)4)56(53-25-11-23-43)55-38-35(54-39(37(38)42)46-24-22-36(44-29(5)48)45-40(46)49)26-52-41(30-12-9-8-10-13-30,31-14-18-33(50-6)19-15-31)32-16-20-34(51-7)21-17-32/h8-10,12-22,24,27-28,35,37-39H,11,25-26H2,1-7H3,(H,44,45,48,49)/t35-,37-,38-,39-,56?/m1/s1
InChI key
CNFKJHKDSRXNFL-UTXREMQHSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Can be employed together with DNA or RNA phosphoramidites
- Recommended deprotection conditions are 8 hours at 55 °C using concentrated ammonia solution, or with AMA for 10 minutes at 65 °C
- Synthesis of 2′Fluoro oligonucleotides is similar to standard DNA synthesis but requires an elongated coupling time (recommended is 3 minutes compared to 90 seconds for DNA monomers)
- Consistent lot-to-lot high purity and performance
- Manufactured under a certified ISO 9001 quality system
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service