Skip to Content
Merck
All Photos(1)

Documents

C0879

Sigma-Aldrich

i-Cholesteryl methyl ether

~95%

Synonym(s):

3α,5-Cyclo-5α-cholestan-6β-ol methyl ether, 6β-Methoxy-3α,5-cyclocholestane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48O
CAS Number:
Molecular Weight:
400.68
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

~95%

storage temp.

−20°C

SMILES string

COC1CC2C3CCC([C@@H](C)CCCC(C)C)C3(C)CCC2C4(C)CC[C@@H]5C[C@@]145

InChI

1S/C28H48O/c1-18(2)8-7-9-19(3)22-10-11-23-21-16-25(29-6)28-17-20(28)12-15-27(28,5)24(21)13-14-26(22,23)4/h18-25H,7-17H2,1-6H3

InChI key

DSVYQBSADVVKNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Do not confuse with cholesteryl methyl ether.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Rao et al.
Journal of steroid biochemistry, 19(4), 1391-1395 (1983-10-01)
A soil microorganism was isolated by the enrichment culture technique using cholesterol as the sole source of carbon. The organism has been identified as belonging to the genus Moraxella. With this organism two novel biotransformations of sterols were observed viz.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service