Skip to Content
Merck
All Photos(1)

Key Documents

C0668

Sigma-Aldrich

Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg

≥90% (HPLC)

Synonym(s):

Laminin Fragment 925-933

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H62N12O14S
CAS Number:
Molecular Weight:
967.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥90% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)CNC(=O)[C@@H]2CCCN2C(=O)[C@H](CC(O)=O)NC(=O)[C@@H](N)CS)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C40H62N12O14S/c1-3-20(2)32(37(63)46-17-30(56)48-27(18-53)35(61)49-24(39(65)66)6-4-12-44-40(42)43)51-34(60)25(14-21-8-10-22(54)11-9-21)47-29(55)16-45-36(62)28-7-5-13-52(28)38(64)26(15-31(57)58)50-33(59)23(41)19-67/h8-11,20,23-28,32,53-54,67H,3-7,12-19,41H2,1-2H3,(H,45,62)(H,46,63)(H,47,55)(H,48,56)(H,49,61)(H,50,59)(H,51,60)(H,57,58)(H,65,66)(H4,42,43,44)/t20-,23-,24-,25-,26-,27-,28-,32-/m0/s1

InChI key

NGBBIWFVAWMQKZ-JDIVNGTKSA-N

Gene Information

human ... POMC(5443)

Amino Acid Sequence

Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg

Biochem/physiol Actions

Inhibits experimental metastasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sofia Silva et al.
International journal of pharmaceutics, 519(1-2), 240-249 (2017-01-24)
Functionalized gold nanoparticles (AuNPs) have been widely investigated as promising multifunctional nanosystems for the theragnosis of lung cancer, the most common and prominent cause of cancer death worldwide. Nevertheless, nanoparticles are not in appropriate sizes for an accurate deep lung
Y Iwamoto et al.
Science (New York, N.Y.), 238(4830), 1132-1134 (1987-11-20)
The invasion of tumor cells through basement membranes is a critical step in the formation of metastases. The binding of the malignant cells to laminin in the basement membranes allows their attachment and activates their invasiveness. Recently a synthetic nonapeptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service