Skip to Content
Merck
All Photos(1)

Key Documents

B9436

Sigma-Aldrich

Monoclonal Anti-Brain-derived Neurotrophic Factor antibody produced in mouse

clone 37141, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-BDNF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

37141, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 1-2 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

Looking for similar products? Visit Product Comparison Guide

General description

Brain derived neurotrophic factor (BDNF) belongs to a family of secreted proteins called neurotrophins that includes, nerve growth factor (NGF), neurotrophin-3 (NT3) and NT4/5. BDNF is expressed in the developing and the mature brain. The influence of BDNF is mediated by its binding to TrkB receptor and sometimes the receptor p75NTR. The downstream pathways promoted by BDNF include PI3K, MAPK and JAK/STAT. The most important role of BDNF is the regulation of synaptic transmission and plasticity, protein synthesis and phosphorylation of local proteins. The secretion of BDNF is highly regulated at both transcriptional and translational levels since it is involved in the proper function of neuronal circuit and development of cognitive functions. The deficits in BDNF functions are implicated in a number of neurodegenerative disorders such as Huntington′s disease, schizophrenia and dementia
Monoclonal Anti-Brain-derived Neurotrophic Factor detects human BDNF.

Immunogen

purified, Sf21-derived, recombinant human brain-derived neurotrophic factor.

Application

Monoclonal Anti-Brain-derived Neurotrophic Factor may be used to detect human BDNF by immunoblotting using a working antibody concentration of 1-2 μg/ml.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth G Pitts et al.
Scientific reports, 8(1), 4495-4495 (2018-03-16)
Specific corticostriatal structures and circuits are important for flexibly shifting between goal-oriented versus habitual behaviors. For example, the orbitofrontal cortex and dorsomedial striatum are critical for goal-directed action, while the dorsolateral striatum supports habits. To determine the role of neurotrophin
Jordi Alberch et al.
Progress in brain research, 146, 195-229 (2004-01-01)
Huntington's disease is a neurodegenerative disorder characterized by the selective loss of striatal neurons and, to a lesser extent, cortical neurons. The neurodegenerative process is caused by the mutation of huntingtin gene. Recent studies have established a link between mutant
Hui Qiao et al.
Brain research, 1663, 29-37 (2017-03-13)
Major depressive disorder (MDD) is one of the most common psychiatric disorder, but the underlying mechanisms are largely unknown. Increasing evidence shows that brain-derived neurotrophic factor (BDNF) plays an important role in the structural plasticity induced by depression. Considering the
Lauren P Shapiro et al.
Journal of neuroscience research, 95(5), 1123-1143 (2016-10-14)
The prevalence of depression, anxiety, schizophrenia, and drug and alcohol use disorders peaks during adolescence. Further, up to 50% of "adult" mental health disorders emerge in adolescence. During adolescence, the prefrontal cortex (PFC) undergoes dramatic structural reorganization, in which dendritic
Mark P Mattson et al.
Trends in neurosciences, 27(10), 589-594 (2004-09-18)
Brain-derived neurotrophic factor (BDNF) and serotonin (5-hydroxytryptamine, 5-HT) are known to regulate synaptic plasticity, neurogenesis and neuronal survival in the adult brain. These two signals co-regulate one another such that 5-HT stimulates the expression of BDNF, and BDNF enhances the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service