Skip to Content
Merck
All Photos(1)

Key Documents

B2801

Millipore

Brucella Agar Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

pH

7.0±0.2 (25 °C)

Application

Recommended for enrichment, isolation and cultivation of the Brucella species.

Other Notes

Components (g/L)
Casein Enzymic Hydrolysate, 10.00
Peptic Digest of Animal Tissue, 10.00
Yeast Extract, 2.00
Dextrose, 1.00
Sodium Chloride, 5.00
Sodium Bisulphite, 0.10
Agar, 15.00

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A selective medium for the isolation of brucellae from contaminated materials.
C D KUZDAS et al.
Journal of bacteriology, 66(4), 502-504 (1953-10-01)
[A selective medium for isolation of Brucella melitensis].
G RENOUX
Annales de l'Institut Pasteur, 87(3), 325-333 (1954-09-01)
A preliminary report on a selective medium for the culture of Brucella, including fastidious types.
L M JONES et al.
Bulletin of the World Health Organization, 19(1), 200-203 (1958-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service