Skip to Content
Merck
All Photos(1)

Key Documents

B2251

Sigma-Aldrich

5-Benzyloxy-DL-tryptophan

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O3
CAS Number:
Molecular Weight:
310.35
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

crystalline

color

light yellow

storage temp.

−20°C

SMILES string

NC(Cc1c[nH]c2ccc(OCc3ccccc3)cc12)C(O)=O

InChI

1S/C18H18N2O3/c19-16(18(21)22)8-13-10-20-17-7-6-14(9-15(13)17)23-11-12-4-2-1-3-5-12/h1-7,9-10,16,20H,8,11,19H2,(H,21,22)

InChI key

DFGNDJBYANKHIO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L T Kushashvili et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 33(1), 16-22 (1988-01-01)
The process of asymmetric deacylation of an acetyl derivative of 5-benzyl hydroxytryptophan racemate in the presence of native microbial aminoacylase was studied. The effect of pH, temperature, concentration of the enzyme and substrate were investigated. Conditions for preparation and isolation
T M Crone et al.
Carcinogenesis, 16(8), 1687-1692 (1995-08-01)
Although the human O6-alkylguanine-DNA alkyltransferase (AGT) is very sensitive to inactivation by O6-benzylguanine (BG) or 2,4-diamino-6-benzyloxy-5-nitrosopyrimidine (5-nitroso-BP), the equivalent protein formed by the carboxyl terminal domain of the product of the Escherichia coli ada gene (Ada-C) is unaffected by these
K Iseki et al.
Biochimica et biophysica acta, 1146(1), 121-126 (1993-02-23)
The uptake mechanisms of organic cations such as tryptamine, tyramine, 5-benzyloxytryptamine (BOTA) and their zwitterionic derivatives (tyrosine, tryptophan, 5-benzyloxytryptophan (BOTP)) by rat intestinal brush-border membrane vesicles and liposome containing phosphatidylserine were studied and compared. As compared to their zwitterionic derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service