Skip to Content
Merck
All Photos(1)

Key Documents

AV32060

Sigma-Aldrich

Anti-PCSK6 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Proprotein convertase subtilisin/kexin type 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

68 kDa

species reactivity

rabbit, dog, horse, human, mouse, pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCSK6(5046)

General description

PCSK6 is a proprotein convertase that processes inactive protein into their active forms. PCSK6 has been linked to handedness in dyslexic individuals and has also been implicated in malignant gliomas.
Rabbit Anti-PCSK6 antibody recognizes human, mouse, rat, and canine PCSK6.

Immunogen

Synthetic peptide directed towards the N terminal region of human PCSK6

Application

Rabbit Anti-PCSK6 antibody can be used for western blot applications at a concentration of 2.5μg/ml.

Biochem/physiol Actions

PCSK6 is a calcium-dependent serine endoprotease that can cleave precursor protein at their paired basic amino acid processing sites. This gene is thought to play a role in tumor progression.

Sequence

Synthetic peptide located within the following region: IYSASWGPDDDGKTVDGPGRLAKQAFEYGIKKGRQGLGSIFVWASGNGGR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Delic et al.
Neuropathology and applied neurobiology, 38(2), 201-212 (2011-07-05)
The molecular mechanisms underlying the infiltrative growth of glioblastomas, the most common primary tumours of the central nervous system in adults, are still poorly understood. We aimed to identify and functionally validate novel glioma invasion-associated candidate genes. Microarray-based expression analysis
Thomas S Scerri et al.
Human molecular genetics, 20(3), 608-614 (2010-11-06)
Approximately 90% of humans are right-handed. Handedness is a heritable trait, yet the genetic basis is not well understood. Here we report a genome-wide association study for a quantitative measure of relative hand skill in individuals with dyslexia [reading disability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service