Skip to Content
Merck
All Photos(1)

Key Documents

A9801

Sigma-Aldrich

1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrapotassium salt

Synonym(s):

BAPTA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C22H20N2O10K4
CAS Number:
Molecular Weight:
628.79
MDL number:
UNSPSC Code:
12352200

solubility

~50 mg/mL (pH >6)

SMILES string

[K].OC(=O)CN(CC(O)=O)c1ccccc1OCCOc2ccccc2N(CC(O)=O)CC(O)=O

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Highly selective calcium chelating reagent.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service