Skip to Content
Merck
All Photos(1)

Key Documents

A9102

Sigma-Aldrich

AH13205

≥98% (HPLC), oil

Synonym(s):

trans-2-(4-(1-Hydroxyhexyl)phenyl-5-oxocyclopentaneheptanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O4
CAS Number:
Molecular Weight:
388.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

oil

color

pale yellow

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

−20°C

SMILES string

CCCCCC(O)c1ccc(cc1)[C@H]2CCC(=O)[C@@H]2CCCCCCC(O)=O

InChI

1S/C24H36O4/c1-2-3-6-10-22(25)19-14-12-18(13-15-19)20-16-17-23(26)21(20)9-7-4-5-8-11-24(27)28/h12-15,20-22,25H,2-11,16-17H2,1H3,(H,27,28)/t20-,21-,22?/m1/s1

InChI key

XMQKDOCUWFCMEJ-JAZPPYFYSA-N

General description

AH13205 is EP2 prostanoid receptor agonist. There are five types of prostanoid receptors, which are classified based on sensitivity to the five naturally-occuring prostanoid ligand. The EP type of receptor is further divided into four subgroups – EP1, EP2, EP3 and EP4. EP2 activation results in vascular, bronchial and reproductive smooth muscle relaxation.

Biochem/physiol Actions

AH13205 is a selective, low potency agonist for the EP2 receptor. AH13205 has been shown to compete with Prostaglandin E2 (PGE2) for binding, resulting in the displacement of PGE2 binding.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K K Meja et al.
British journal of pharmacology, 122(1), 149-157 (1997-09-23)
1. The prostanoid receptor(s) that mediates inhibition of bacterial lipopolysaccharide (LPS)-induced tumour necrosis factor-alpha (TNF-alpha) generation from human peripheral blood monocytes was classified by use of naturally occurring and synthetic prostanoid agonists and antagonists. 2. In human monocytes that were
G C Smith et al.
The Journal of pharmacology and experimental therapeutics, 271(1), 390-396 (1994-10-01)
The purpose of this study was to determine which receptors mediate the dilator effects of prostaglandins (PGs) on the ductus arteriosus of the fetal rabbit. Isolated rings of the vessel from fetal New Zealand White rabbits were precontracted with indomethacin
D F Woodward et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 11(3), 447-454 (1995-01-01)
The cloning of the genes that encode for prostaglandin (PG) receptors has resolved much of the complexity and controversy in this area by confirming the classification proposed by Coleman, et al. Two issues that remained unresolved were (1) the inability
S A Milne et al.
Prostaglandins, 49(4), 225-237 (1995-04-01)
A fourth PGE receptor subtype, the EP4 receptor, has recently been described in the pig saphenous vein (PSV). Similar to the EP2 receptor, it mediates relaxation and is linked to stimulation of adenylate cyclase. The aim of this study was
S J Lydford et al.
British journal of pharmacology, 112(1), 133-136 (1994-05-01)
1. Using a range of natural and synthetic prostanoid receptor agonists and antagonists, we have shown that the rat isolated trachea contains a heterogeneous population of prostaglandin receptor sub-types mediating both relaxation and contraction of the smooth muscle. Prostaglandin E2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service