Skip to Content
Merck
All Photos(1)

Key Documents

A8889

Sigma-Aldrich

N6-([6-Aminohexyl]carbamoylmethyl)adenosine 5′-triphosphate lithium salt

≥85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32N7O14P3 · xLi+
CAS Number:
Molecular Weight:
663.41 (free acid basis)
UNSPSC Code:
12352202

Assay

≥85%

storage temp.

−20°C

Application

Side-chain functionalized derivative of ATP, useful for preparation of immobilized ATP such as for affinity purifications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Lindberg et al.
European journal of biochemistry, 53(2), 481-486 (1975-05-06)
Alkylation of ATP with iodoacetic acid at pH 6.5 yielded 1-carboxymethyl-ATP which, after alkaline rearrangement, gave N-6-carboxymethyl-ATP. Condensation of this analogue with 1,6-diaminohexane in the presence of a water-soluble carbodiimide generated N-6-[(6-aminohexyl)carbamoylmethyl]-ATP in an overall yield of 40% based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service