A6472
14-Acetylvirescenine from Delphinium sp.
≥99% (TLC), white, powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C25H39NO7
Molecular Weight:
465.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥99% (TLC)
form
powder
color
white
storage temp.
2-8°C
SMILES string
COC[C@@]12CC[C@H](O)[C@@]34C5C[C@@H]6[C@H](C[C@@](O)(C5[C@H]6OC(C)=O)[C@@](O)(CC13)C4N(CO)C2)OC
Application
14-Acetylvirescenine blocks action potential propagation at the nicotinic receptors of vegetative ganglia and neuromuscular junctions. 14-Acetylvirescenine is hypotensive with curare-like activity.
Biochem/physiol Actions
Blocks the propagation of the action potential at the nicotinic cholinoreceptors of vegetative ganglia and neuro-muscular junctions. Hypotensive and curare-like action, I.V. and orally.
Warning
Protect from light.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service