Skip to Content
Merck
All Photos(1)

Documents

A4469

Sigma-Aldrich

Monoclonal Anti-ACTR/AIB1 antibody produced in mouse

clone AX 15, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

AX 15, monoclonal

mol wt

antigen 150 kDa

contains

0.035% sodium azide
30% glycerol

species reactivity

human

technique(s)

immunoprecipitation (IP): 15 μL using 500 μg of a BG-1 ovarian cell RIPA lysate.
western blot (chemiluminescent): 1:500 using RIPA lysates of BG-1 ovarian cancer cells

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCOA3(8202)

General description

ACTR associates with estrogen receptors in breast and ovarian cancers and stimulates transcription. Hence, ACTR has been implicated in the carcinogenesis of breast and ovarian tumors. Mouse monoclonal Anti-ACTR/AIB1 antibody recognizes human ACTR (150kD) and another doublet (110kD) by immunoblotting.
ACTR, a nuclear receptor coactivator, is also referred to as amplified in breast cancer 1 (AIB1), thyroid hormone receptor activator molecule-1 (TRAM-1), receptor-associated coactivator 3 (RAC3 and SRC3).

Immunogen

GST fusion protein containing amino acids 605-1294 of human ACTR.

Application

Mouse monoclonal can be used for immunoprecipitation (15μL using 500μg of a BG-1 ovarian cell RIPA lysate) and western blot (1:500, using RIPA lysates of BG-1 ovarian cancer cells) assays.

Physical form

The product is supplied as mouse ascites fluid, containing 30% glycerol and 0.035% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Anzick et al.
Science (New York, N.Y.), 277(5328), 965-968 (1997-08-15)
Members of the recently recognized SRC-1 family of transcriptional coactivators interact with steroid hormone receptors to enhance ligand-dependent transcription. AIB1, a member of the SRC-1 family, was cloned during a search on the long arm of chromosome 20 for genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service